Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001551.3 | NP_001001551.2 | 5 - 5 | 867 | 147 |
NM_001256915.1 | NP_001243844.1 | 5 - 5 | 1041 | 147 | |
NR_046421.1 | 4 - 4 | 836 | 147 | ||
NR_046422.1 | 6 - 6 | 1453 | 147 | ||
XM_006717110.3 | XP_006717173.1 | 5 - 5 | 986 | 147 | |
XM_006717111.3 | XP_006717174.1 | 5 - 5 | 3395 | 147 | |
XM_017014719.1 | XP_016870208.1 | 3 - 3 | 792 | 147 | |
XM_017014724.1 | XP_016870213.1 | 5 - 5 | 1698 | 147 | |
XM_017014725.1 | XP_016870214.1 | 5 - 5 | 827 | 147 | |
GenBank mRNA | AF086281.1 | - | 1 - 1 | 444 | 147 |
AY615889.1 | - | 5 - 5 | 758 | 147 | |
BC036421.1 | - | 6 - 6 | 1456 | 147 | |
BC045782.1 | - | 6 - 6 | 1451 | 147 | |
BC142991.1 | - | 6 - 6 | 958 | 147 | |
W69675.1 | - | 1 - 1 | 147 |