Assay Design



Gene Transcripts

Gene Symbol
OR4C46
Entrez Gene ID
119749
Gene Name
olfactory receptor family 4 subfamily C member 46
Gene Aliases
-
Chromosome Location
Chr.11: 54603069 - 54603998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004703.1 NP_001004703.1 1 - 1 639 155

View Full Product Details