Assay Design



Gene Transcripts

Gene Symbol
OR4A16
Entrez Gene ID
81327
Gene Name
olfactory receptor family 4 subfamily A member 16
Gene Aliases
OR11-117, OR4A16Q
Chromosome Location
Chr.11: 55343201 - 55344187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005274.1 NP_001005274.1 1 - 1 936 127
GenBank mRNA BC140748.1 - 1 - 1 986 127

View Full Product Details