Assay Design
Gene Transcripts
- Gene Symbol
- HLA-L
- Entrez Gene ID
-
3139
- Gene Name
- major histocompatibility complex, class I, L (pseudogene)
- Gene Aliases
-
HLA-92, HLA92, HLAL
- Chromosome Location
-
Chr.6: 30259562 - 30266951 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027822.1 |
|
5 - 6 |
900 |
136 |
GenBank mRNA |
AK127349.1 |
- |
5 - 6 |
865 |
136 |
BC139904.1 |
- |
4 - 5 |
685 |
136 |
View Full Product Details