Assay Design



Gene Transcripts

Gene Symbol
GK
Entrez Gene ID
2710
Gene Name
glycerol kinase
Gene Aliases
GK1, GKD
Chromosome Location
Chr.X: 30653348 - 30731462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000167.5 NP_000158.1 6 - 7 734 143
NM_001128127.2 NP_001121599.1 6 - 7 734 143
NM_001205019.1 NP_001191948.1 6 - 7 734 143
NM_203391.3 NP_976325.1 6 - 7 734 143
XM_005274488.4 XP_005274545.1 4 - 5 504 143
XM_006724483.2 XP_006724546.1 7 - 8 771 143
XM_006724484.2 XP_006724547.1 7 - 8 770 143
XM_006724485.2 XP_006724548.1 5 - 6 579 143
XM_006724486.3 XP_006724549.1 4 - 5 564 143
XM_011545491.2 XP_011543793.1 7 - 8 772 143
XM_011545492.1 XP_011543794.1 7 - 8 771 143
XM_011545493.2 XP_011543795.1 5 - 6 454 143
XM_011545494.2 XP_011543796.1 5 - 6 399 143
XM_017029409.1 XP_016884898.1 7 - 8 769 143
XM_017029410.1 XP_016884899.1 4 - 5 548 143
XM_017029411.1 XP_016884900.1 5 - 6 581 143
XM_017029412.1 XP_016884901.1 4 - 5 502 143
GenBank mRNA AK294938.1 - 7 - 8 769 143
AK308238.1 - 4 - 5 413 143
AK313215.1 - 6 - 7 670 143
BC037549.1 - 6 - 7 591 143
BC042421.1 - 6 - 7 735 143
BC071595.1 - 6 - 7 735 143
BX647604.1 - 6 - 7 555 143
L13943.1 - 6 - 7 621 143
X69886.1 - 2 - 3 170 143

View Full Product Details