Assay Design



Gene Transcripts

Gene Symbol
OR6C6
Entrez Gene ID
283365
Gene Name
olfactory receptor family 6 subfamily C member 6
Gene Aliases
-
Chromosome Location
Chr.12: 55294288 - 55295232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005493.1 NP_001005493.1 1 - 1 718 146

View Full Product Details