Assay Design



Gene Transcripts

Gene Symbol
NMS
Entrez Gene ID
129521
Gene Name
neuromedin S
Gene Aliases
-
Chromosome Location
Chr.2: 100470482 - 100483280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011717.1 NP_001011717.1 3 - 4 187 167
GenBank mRNA AB164464.1 - 3 - 4 187 167

View Full Product Details