Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201628.2 | NP_963922.2 | 11 - 12 | 2065 | 139 |
XM_005245795.4 | XP_005245852.1 | 13 - 14 | 2320 | 139 | |
XM_011541074.2 | XP_011539376.1 | 13 - 14 | 2327 | 139 | |
XM_011541075.2 | XP_011539377.1 | 11 - 12 | 1983 | 139 | |
XM_011541076.2 | XP_011539378.1 | 11 - 12 | 2128 | 139 | |
XM_011541077.2 | XP_011539379.1 | 11 - 12 | 3107 | 139 | |
XM_011541081.2 | XP_011539383.1 | 5 - 6 | 807 | 139 | |
GenBank mRNA | AK293806.1 | - | 6 - 7 | 871 | 139 |
BC101637.1 | - | 1 - 2 | 146 | 139 | |
BC113621.1 | - | 1 - 2 | 146 | 139 | |
DA317442.1 | - | 1 - 2 | 83 | 139 | |
DN831635.1 | - | 2 - 3 | 347 | 139 |