Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FHOD1 | NM_001318202.1 | NP_001305131.1 | ||
NM_013241.2 | NP_037373.2 | |||
XM_006721180.1 | XP_006721243.1 | |||
XM_011523043.2 | XP_011521345.1 | |||
XM_011523044.1 | XP_011521346.1 | |||
XM_011523045.2 | XP_011521347.1 | |||
AB041046.1 | BAD06250.1 | |||
AB209584.1 | BAD92821.1 | |||
AF113615.1 | AAD39906.1 | |||
AK025992.1 | ||||
AK301157.1 | ||||
AY192154.1 | AAO38757.1 | |||
BC033084.1 | AAH33084.1 | |||
BU689676.1 | ||||
KJ658417.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr16:67153308 - 68071571 on Build GRCh38 | Gain | LOC105369155 DDX28 LOC100131303 CARMIL2 SLC9A5 RANBP10 LCAT NRN1L PSKH1 TPPP3 KCTD19 NUTF2 KIAA0895L LRRC36 GFOD2 LRRC29 DPEP2 ACD AGRP PLEKHG4 ENKD1 ELMO3 SLC12A4 THAP11 PARD6A EDC4 CENPT DPEP3 HSF4 MIR328 TRADD ZDHHC1 E2F4 CTRL NOL3 ATP6V0D1 CTCF EXOC3L1 PSMB10 TSNAXIP1 C16orf86 FAM65A FBXL8 DUS2 TMEM208 LOC107984813 LOC100505942 FHOD1 HSD11B2 |
nsv827707 | Chr16:67148355 - 67289296 on Build GRCh38 | Loss | LOC105369155 ELMO3 EXOC3L1 FBXL8 TMEM208 KIAA0895L HSF4 B3GNT9 SLC9A5 MIR328 FHOD1 TRADD E2F4 LRRC29 NOL3 PLEKHG4 C16orf70 |
nsv952044 | Chr16:67141198 - 67244397 on Build GRCh38 | Deletion | LOC105369155 ELMO3 EXOC3L1 FBXL8 TMEM208 KIAA0895L HSF4 B3GNT9 MIR328 FHOD1 TRADD E2F4 LRRC29 NOL3 C16orf70 |
nsv457513 | Chr16:67181417 - 67282697 on Build GRCh38 | Loss | LOC105369155 ELMO3 FHOD1 EXOC3L1 E2F4 LRRC29 TMEM208 KIAA0895L PLEKHG4 SLC9A5 MIR328 |
nsv524492 | Chr16:67174030 - 67294550 on Build GRCh38 | Loss | LOC105369155 ELMO3 EXOC3L1 TMEM208 KIAA0895L SLC9A5 MIR328 FHOD1 E2F4 LRRC29 NOL3 PLEKHG4 KCTD19 |
nsv524363 | Chr16:67136529 - 68071571 on Build GRCh38 | Loss | LOC105369155 DDX28 LOC100131303 CARMIL2 SLC9A5 RANBP10 LCAT NRN1L PSKH1 TPPP3 KCTD19 C16orf70 NUTF2 KIAA0895L LRRC36 GFOD2 B3GNT9 LRRC29 DPEP2 ACD AGRP PLEKHG4 ENKD1 ELMO3 SLC12A4 THAP11 PARD6A EDC4 CENPT DPEP3 HSF4 MIR328 TRADD ZDHHC1 E2F4 CTRL NOL3 ATP6V0D1 CTCF EXOC3L1 PSMB10 TSNAXIP1 C16orf86 FAM65A FBXL8 DUS2 TMEM208 LOC107984813 LOC100505942 FHOD1 HSD11B2 |