Assay Design



Gene Transcripts

Gene Symbol
GOLGA6L1
Entrez Gene ID
283767
Gene Name
golgin A6 family-like 1
Gene Aliases
-
Chromosome Location
Chr.15: 23127066 - 23136839 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001413.3 NP_001001413.3 4 - 5 567 70
XM_017022085.1 XP_016877574.1 4 - 5 584 70
GenBank mRNA AK097517.1 - 4 - 5 549 70
BC127816.1 - 4 - 5 351 70

Gene Symbol
GOLGA6L22
Entrez Gene ID
440243
Gene Name
golgin A6 family-like 22
Gene Aliases
-
Chromosome Location
Chr.15: 22458903 - 22468026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271664.2 NP_001258593.2 5 - 6 509 70
XM_011543882.1 XP_011542184.1 4 - 5 549 70

Gene Symbol
GOLGA6L6
Entrez Gene ID
727832
Gene Name
golgin A6 family-like 6
Gene Aliases
-
Chromosome Location
Chr.15: 20531856 - 20541800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145004.2 NP_001138476.2 4 - 5 531 70

Gene Symbol
LOC100132202
Entrez Gene ID
100132202
Gene Name
golgin subfamily A member 6-like protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 28549682 - 28561099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017022770.1 XP_016878259.1 4 - 5 371 70

Gene Symbol
LOC102723623
Entrez Gene ID
102723623
Gene Name
golgin subfamily A member 6-like protein 6
Gene Aliases
-
Chromosome Location
Chr.15: 23324535 - 23334306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720795.3 XP_006720858.1 4 - 5 565 70
XM_017022778.1 XP_016878267.1 4 - 5 569 70

View Full Product Details