Assay Design
Gene Transcripts
- Gene Symbol
- MESP2
- Entrez Gene ID
-
145873
- Gene Name
- mesoderm posterior bHLH transcription factor 2
- Gene Aliases
-
SCDO2, bHLHc6
- Chromosome Location
-
Chr.15: 89776358 - 89778754 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL360139.1 |
- |
2 - 3 |
168 |
171 |
View Full Product Details