Product Details

Assay Reference Genome
Location

Chr.6:43613675 on build GRCh38
Cytoband
6p21.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5429
Gene Symbol
POLH
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
Location
Chr.6:43576141-43620523 on build GRCh38
Assay Gene Location
Overlaps Intron 10 - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
POLH NM_001291969.1 NP_001278898.1
NM_001291970.1 NP_001278899.1
NM_006502.2 NP_006493.1
XM_005249186.3 XP_005249243.1
XM_011514698.2 XP_011513000.1
AB024313.1 BAA81666.1
AF158185.1 AAD43810.1
AJ420484.1
AK023893.1
AK122807.1
AK294430.1
AK307660.1
AK316151.1
BC015742.1 AAH15742.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv5281 Chr6:43611090 - 43655905 on Build GRCh38 Deletion POLH RSPH9 MAD2L1BP GTPBP2

View Full Product Details