Assay Design
Gene Transcripts
- Gene Symbol
- SULT2B1
- Entrez Gene ID
-
6820
- Gene Name
- sulfotransferase family 2B member 1
- Gene Aliases
-
HSST2
- Chromosome Location
-
Chr.19: 48552172 - 48599427 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_004605.2 |
NP_004596.2 |
1 - 1 |
221 |
90 |
GenBank mRNA |
U92314.1 |
- |
1 - 1 |
417 |
90 |
View Full Product Details