Assay Design



Gene Transcripts

Gene Symbol
SLC15A1
Entrez Gene ID
6564
Gene Name
solute carrier family 15 member 1
Gene Aliases
HPECT1, HPEPT1, PEPT1
Chromosome Location
Chr.13: 98683801 - 98752675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005073.3 NP_005064.1 23 - 23 2991 113
GenBank mRNA AW449979.1 - 1 - 1 113
U21936.1 - 23 - 23 2967 113

View Full Product Details