Assay Design
Gene Transcripts
- Gene Symbol
- SLCO1A2
- Entrez Gene ID
-
6579
- Gene Name
- solute carrier organic anion transporter family member 1A2
- Gene Aliases
-
OATP, OATP-A, OATP1A2, SLC21A3
- Chromosome Location
-
Chr.12: 21264600 - 21403657 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF085224.1 |
- |
13 - 13 |
2250 |
106 |
View Full Product Details