Assay Design



Gene Transcripts

Gene Symbol
SULT1E1
Entrez Gene ID
6783
Gene Name
sulfotransferase family 1E member 1
Gene Aliases
EST, EST-1, ST1E1, STE
Chromosome Location
Chr.4: 69841212 - 69860152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005420.2 NP_005411.1 7 - 7 771 100
GenBank mRNA AK312236.1 - 6 - 6 670 100
BC027956.1 - 7 - 7 771 100
CR407621.1 - 6 - 6 658 100
S77383.1 - 7 - 7 694 100
U08098.1 - 7 - 7 764 100
U55764.1 - 2 - 2 117 100
Y11195.1 - 7 - 7 720 100

View Full Product Details