Assay Design



Gene Transcripts

Gene Symbol
CYP4F8
Entrez Gene ID
11283
Gene Name
cytochrome P450 family 4 subfamily F member 8
Gene Aliases
CPF8, CYPIVF8
Chromosome Location
Chr.19: 15615218 - 15629638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007253.3 NP_009184.1 13 - 13 1716 136
GenBank mRNA AI401776.1 - 1 - 1 136
AK092612.1 - 2 - 2 136
DB329701.1 - 1 - 1 136

View Full Product Details