Assay Design
Gene Transcripts
- Gene Symbol
- FAM74A4
- Entrez Gene ID
-
401508
- Gene Name
- family with sequence similarity 74 member A4
- Gene Aliases
-
FAM74A2
- Chromosome Location
-
Chr.9: 61205260 - 61212373 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026802.1 |
|
1 - 1 |
578 |
154 |
NR_110998.1 |
|
1 - 1 |
578 |
154 |
GenBank mRNA |
AK127145.1 |
- |
1 - 1 |
440 |
154 |
BC121813.1 |
- |
1 - 1 |
432 |
154 |
- Gene Symbol
- FAM74A3
- Entrez Gene ID
-
728495
- Gene Name
- family with sequence similarity 74 member A3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 66970162 - 66977317 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026801.1 |
|
1 - 1 |
606 |
154 |
- Gene Symbol
- FAM74A6
- Entrez Gene ID
-
653123
- Gene Name
- family with sequence similarity 74 member A6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 60929782 - 60935727 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110999.1 |
|
1 - 1 |
432 |
154 |
View Full Product Details