Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002916.4 | NP_001002916.3 | 1 - 1 | 258 | 89 |
GenBank mRNA | AY283370.1 | - | 1 - 1 | 231 | 89 |
BC038109.2 | - | 1 - 1 | 255 | 89 | |
BC118604.1 | - | 1 - 1 | 251 | 89 | |
BC121816.1 | - | 1 - 1 | 251 | 89 |