Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006120.3 | NP_006111.2 | 3 - 3 | 500 | 93 |
GenBank mRNA | AK291858.1 | - | 3 - 3 | 436 | 93 |
AK298081.1 | - | 3 - 3 | 1078 | 93 | |
BC011447.1 | - | 3 - 3 | 473 | 93 | |
BC026279.1 | - | 3 - 3 | 429 | 93 | |
CR450324.1 | - | 3 - 3 | 414 | 93 | |
X62744.1 | - | 3 - 3 | 459 | 93 |