Assay Design



Gene Transcripts

Gene Symbol
CBX8
Entrez Gene ID
57332
Gene Name
chromobox 8
Gene Aliases
PC3, RC1
Chromosome Location
Chr.17: 79794377 - 79797116 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020649.2 NP_065700.1 5 - 5 1379 110
GenBank mRNA AF174482.1 - 5 - 5 1354 110
AF266479.2 - 5 - 5 1310 110
AK074560.1 - 5 - 5 1379 110
BC008937.2 - 5 - 5 1276 110
BC009376.2 - 5 - 5 1332 110
BC019289.1 - 5 - 5 1276 110

View Full Product Details