Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021134.3 | NP_066957.3 | 5 - 5 | 683 | 110 |
GenBank mRNA | BC027710.2 | - | 5 - 5 | 598 | 110 |
BM670857.1 | - | 1 - 1 | 110 | ||
U26596.1 | - | 5 - 5 | 646 | 110 | |
Z49254.1 | - | 5 - 5 | 646 | 110 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006724896.2 | XP_006724959.1 | 5 - 5 | 683 | 110 |