Assay Design



Gene Transcripts

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location
Chr.16: 53054991 - 53327502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 38 - 38 10778 97
NM_025134.4 NP_079410.4 39 - 39 10775 97
XM_005256168.3 XP_005256225.1 39 - 39 10868 97
XM_005256169.2 XP_005256226.1 39 - 39 10890 97
XM_005256172.3 XP_005256229.1 39 - 39 10756 97
XM_005256176.3 XP_005256233.1 38 - 38 9475 97
XM_006721281.2 XP_006721344.1 39 - 39 10807 97
XM_006721283.3 XP_006721346.1 39 - 39 11028 97
XM_011523347.2 XP_011521649.1 40 - 40 10940 97
XM_017023718.1 XP_016879207.1 40 - 40 11063 97
XM_017023719.1 XP_016879208.1 41 - 41 11014 97
XM_017023720.1 XP_016879209.1 40 - 40 10908 97
XM_017023721.1 XP_016879210.1 40 - 40 11061 97
XM_017023723.1 XP_016879212.1 40 - 40 11013 97
XM_017023724.1 XP_016879213.1 40 - 40 10890 97
XM_017023725.1 XP_016879214.1 40 - 40 10842 97
XM_017023726.1 XP_016879215.1 38 - 38 10097 97
XM_017023727.1 XP_016879216.1 38 - 38 9427 97
XM_017023728.1 XP_016879217.1 40 - 40 10924 97
GenBank mRNA AB002306.3 - 38 - 38 10201 97
AY243500.1 - 38 - 38 10701 97
AY243504.1 - 1 - 1 3218 97
AY647157.1 - 39 - 39 10775 97
DQ059482.1 - 29 - 29 8103 97
DQ333316.1 - 39 - 39 10869 97

View Full Product Details