Assay Design



Gene Transcripts

Gene Symbol
APOBEC3A
Entrez Gene ID
200315
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3A
Gene Aliases
A3A, ARP3, PHRBN, bK150C2.1
Chromosome Location
Chr.22: 38957522 - 38963183 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270406.1 NP_001257335.1 5 - 5 1344 166
NM_145699.3 NP_663745.1 5 - 5 1398 166

View Full Product Details