Assay Design



Gene Transcripts

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
Chromosome Location
Chr.6: 26365159 - 26451697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197246.2 NP_001184175.1 6 - 7 972 108
NM_001197247.2 NP_001184176.1 8 - 9 1237 108
NM_001197248.2 NP_001184177.1 6 - 7 1076 108
NM_001197249.2 NP_001184178.1 7 - 8 1137 108
NM_007047.4 NP_008978.2 8 - 9 1227 108
XM_005248827.3 XP_005248884.1 8 - 9 1215 108
XM_006714979.3 XP_006715042.1 8 - 9 1230 108
XM_006714980.3 XP_006715043.1 8 - 9 1230 108
XM_006714981.3 XP_006715044.1 8 - 9 1220 108
XM_006714982.3 XP_006715045.1 7 - 8 1140 108
XM_011514267.2 XP_011512569.1 8 - 9 1096 108
XM_011514268.2 XP_011512570.1 8 - 9 1230 108
XM_011514269.2 XP_011512571.1 7 - 8 999 108
XM_011514270.2 XP_011512572.1 7 - 8 933 108
XM_011514271.2 XP_011512573.1 7 - 8 799 108
XM_017010211.1 XP_016865700.1 8 - 9 1205 108
XM_017010212.1 XP_016865701.1 8 - 9 1081 108
XM_017010213.1 XP_016865702.1 7 - 8 1102 108
XM_017010214.1 XP_016865703.1 7 - 8 1125 108
XM_017010215.1 XP_016865704.1 7 - 8 933 108
XM_017010217.1 XP_016865706.1 6 - 7 843 108
GenBank mRNA AF257505.1 - 6 - 7 974 108
AK290880.1 - 8 - 9 1165 108
AK299417.1 - 7 - 8 1021 108
AK303600.1 - 6 - 7 993 108
BC002832.2 - 8 - 9 1118 108
BC020214.1 - 8 - 9 1118 108
U90144.1 - 7 - 8 1034 108
U90546.1 - 8 - 9 1154 107

View Full Product Details