Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300850.1 | NP_001287779.1 | 9 - 10 | 1704 | 126 |
NM_001300851.1 | NP_001287780.1 | 9 - 10 | 1704 | 126 | |
NM_001300852.1 | NP_001287781.1 | 9 - 10 | 1704 | 126 | |
NM_172071.3 | NP_742068.1 | 9 - 10 | 1704 | 126 | |
XM_011509231.2 | XP_011507533.1 | 9 - 10 | 1704 | 126 | |
GenBank mRNA | AB095945.1 | - | 9 - 10 | 1545 | 126 |
AK122948.1 | - | 9 - 10 | 1704 | 126 | |
BC136784.1 | - | 9 - 10 | 1658 | 126 | |
BC144408.1 | - | 9 - 10 | 1658 | 126 |