Assay Design



Gene Transcripts

Gene Symbol
OTUD1
Entrez Gene ID
220213
Gene Name
OTU deubiquitinase 1
Gene Aliases
DUBA7, OTDC1
Chromosome Location
Chr.10: 23439269 - 23442381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145373.2 NP_001138845.1 1 - 1 1401 93
GenBank mRNA AB188491.1 - 1 - 1 1401 93
AK096389.1 - 1 - 1 246 93

View Full Product Details