Assay Design
Gene Transcripts
- Gene Symbol
- CFAP36
- Entrez Gene ID
-
112942
- Gene Name
- cilia and flagella associated protein 36
- Gene Aliases
-
BARTL1, CCDC104
- Chromosome Location
-
Chr.2: 55519595 - 55545080 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC001797.1 |
- |
- |
2127 |
150 |
- Gene Symbol
- MT-ND4
- Entrez Gene ID
-
4538
- Gene Name
- mitochondrially encoded NADH dehydrogenase 4
- Gene Aliases
-
MTND4, ND4
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
NC_012920.ND4.0 |
- |
- |
574 |
150 |
- Gene Symbol
- HNRNPM
- Entrez Gene ID
-
4670
- Gene Name
- heterogeneous nuclear ribonucleoprotein M
- Gene Aliases
-
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M
- Chromosome Location
-
Chr.19: 8444575 - 8489118 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK309659.1 |
- |
- |
1358 |
150 |
- Gene Symbol
- ZNF232
- Entrez Gene ID
-
7775
- Gene Name
- zinc finger protein 232
- Gene Aliases
-
ZSCAN11
- Chromosome Location
-
Chr.17: 5105734 - 5123116 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC002852.1 |
- |
- |
2501 |
150 |
View Full Product Details