Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022831.2 | NP_073742.2 | 5 - 6 | 556 | 150 |
GenBank mRNA | AK022868.1 | - | 5 - 6 | 508 | 150 |
AK289865.1 | - | 5 - 6 | 507 | 150 | |
AK301870.1 | - | 5 - 6 | 682 | 150 | |
AL833718.1 | - | 5 - 6 | 346 | 150 | |
BC015535.1 | - | 5 - 6 | 483 | 150 | |
BC043142.1 | - | 5 - 6 | 1604 | 150 | |
BC067805.1 | - | 5 - 6 | 504 | 150 | |
BC171848.1 | - | 5 - 6 | 531 | 150 |