Assay Design



Gene Transcripts

Gene Symbol
SMR3A
Entrez Gene ID
26952
Gene Name
submaxillary gland androgen regulated protein 3A
Gene Aliases
P-B1, PBI, PRL5, PROL5
Chromosome Location
Chr.4: 70360776 - 70367158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012390.3 NP_036522.3 3 - 3 451 160
XM_017008021.1 XP_016863510.1 2 - 2 422 160
GenBank mRNA BC140927.1 - 2 - 2 397 160
CD722957.1 - 3 - 3 451 160

View Full Product Details