Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040433.1 | NP_001035523.1 | 1 - 2 | 109 | 74 |
GenBank mRNA | AY396740.1 | - | 1 - 2 | 109 | 74 |
BC137530.1 | - | 1 - 2 | 80 | 74 | |
BC137531.1 | - | 1 - 2 | 80 | 74 |