Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002289.2 | NP_002280.1 | 3 - 4 | 394 | 96 |
XM_006719395.3 | XP_006719458.1 | 4 - 5 | 472 | 96 | |
GenBank mRNA | BC069103.1 | - | 3 - 4 | 394 | 96 |
BC112316.1 | - | 3 - 4 | 390 | 96 | |
BC112318.1 | - | 3 - 4 | 390 | 96 | |
BX281629.1 | - | 1 - 2 | 70 | 96 | |
CR541987.1 | - | 3 - 4 | 368 | 96 | |
CR542017.1 | - | 3 - 4 | 368 | 96 | |
J00270.1 | - | 3 - 4 | 368 | 96 |