Assay Design



Gene Transcripts

Gene Symbol
SLC16A5
Entrez Gene ID
9121
Gene Name
solute carrier family 16 member 5
Gene Aliases
MCT5, MCT6
Chromosome Location
Chr.17: 75087727 - 75106590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004695.3 NP_004686.1 1 - 2 100 68
XM_017025293.1 XP_016880782.1 1 - 2 78 68
GenBank mRNA BC009684.1 - 1 - 2 81 68
BI820515.1 - 1 - 2 100 68

View Full Product Details