Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159547.1 | NP_001153019.1 | 2 - 3 | 1066 | 52 |
NM_207406.3 | NP_997289.2 | 2 - 3 | 1066 | 52 | |
XM_017008185.1 | XP_016863674.1 | 1 - 2 | 723 | 52 | |
GenBank mRNA | AB126828.1 | - | 2 - 3 | 1066 | 52 |
AB126829.1 | - | 2 - 3 | 1066 | 52 | |
AK125953.1 | - | 2 - 3 | 252 | 52 |