Assay Design
Gene Transcripts
- Gene Symbol
- HIST2H3D
- Entrez Gene ID
-
653604
- Gene Name
- histone cluster 2, H3d
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 149813225 - 149813681 on Build GRCh38
- Gene Symbol
- HIST2H3A
- Entrez Gene ID
-
333932
- Gene Name
- histone cluster 2, H3a
- Gene Aliases
-
H3/n, H3/o
- Chromosome Location
-
Chr.1: 149852619 - 149853125 on Build GRCh38
- Gene Symbol
- HIST2H3C
- Entrez Gene ID
-
126961
- Gene Name
- histone cluster 2, H3c
- Gene Aliases
-
H3, H3.2, H3/M, H3F2, H3FM, H3FN
- Chromosome Location
-
Chr.1: 149840687 - 149841193 on Build GRCh38
View Full Product Details