Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077619.1 | NP_001071087.1 | 6 - 7 | 800 | 71 |
GenBank mRNA | AK126300.1 | - | 6 - 7 | 703 | 71 |
AL831990.2 | - | 6 - 7 | 674 | 71 | |
AL833075.2 | - | 6 - 7 | 674 | 71 | |
BC113645.1 | - | 6 - 7 | 800 | 71 | |
BX647876.2 | - | 6 - 7 | 674 | 71 |