Assay Design
Gene Transcripts
- Gene Symbol
- IFNA16
- Entrez Gene ID
-
3449
- Gene Name
- interferon alpha 16
- Gene Aliases
-
IFN-alpha-16, IFN-alphaO
- Chromosome Location
-
Chr.9: 21216373 - 21217311 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_002173.3 |
NP_002164.1 |
1 - 1 |
857 |
155 |
GenBank mRNA |
M28585.1 |
- |
1 - 1 |
857 |
155 |
View Full Product Details