Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004610.3 | NP_004601.3 | 7 - 8 | 999 | 73 |
XM_011536082.2 | XP_011534384.1 | 5 - 6 | 904 | 73 | |
XM_011536084.2 | XP_011534386.1 | 7 - 8 | 1003 | 73 | |
XM_011536085.2 | XP_011534387.1 | 7 - 8 | 1182 | 73 | |
XM_011536088.2 | XP_011534390.1 | 6 - 7 | 822 | 73 | |
XM_011536090.2 | XP_011534392.1 | 6 - 7 | 769 | 73 |