Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004232.3 | NP_004223.2 | 2 - 2 | 1151 | 161 |
XM_005266783.3 | XP_005266840.1 | 2 - 2 | 1113 | 161 | |
XM_017026086.1 | XP_016881575.1 | 2 - 2 | 1157 | 161 | |
XM_017026087.1 | XP_016881576.1 | 2 - 2 | 1090 | 161 | |
GenBank mRNA | AB006968.1 | - | 1 - 1 | 835 | 161 |
BC020082.1 | - | 2 - 2 | 1025 | 161 |