Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287214.1 | NP_001274143.1 | 3 - 4 | 660 | 144 |
NM_001287215.1 | NP_001274144.1 | 4 - 5 | 510 | 144 | |
NM_001634.5 | NP_001625.2 | 4 - 5 | 747 | 144 | |
GenBank mRNA | AK300330.1 | - | 3 - 4 | 659 | 144 |
AK304351.1 | - | 4 - 5 | 510 | 144 | |
AK316471.1 | - | 4 - 5 | 379 | 144 | |
BC000171.2 | - | 4 - 5 | 520 | 144 | |
BX640599.1 | - | 4 - 5 | 747 | 144 | |
M21154.1 | - | 4 - 5 | 144 |