Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138815.3 | NP_620170.3 | 8 - 9 | 1173 | 99 |
XM_011512443.1 | XP_011510745.1 | 8 - 9 | 1014 | 99 | |
XM_011512444.2 | XP_011510746.1 | 8 - 9 | 1156 | 99 | |
XM_011512445.2 | XP_011510747.1 | 8 - 9 | 1039 | 99 | |
GenBank mRNA | AY283672.1 | - | 8 - 9 | 1175 | 99 |
BC018070.1 | - | 8 - 9 | 1051 | 99 |