Assay Design



Gene Transcripts

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
Chromosome Location
Chr.11: 123057492 - 123062366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153201.3 NP_694881.1 7 - 8 1664 87
GenBank mRNA AB034951.1 - 7 - 8 1428 87
AK222628.1 - 7 - 8 1462 87

View Full Product Details