Assay Design



Gene Transcripts

Gene Symbol
RPS25
Entrez Gene ID
6230
Gene Name
ribosomal protein S25
Gene Aliases
S25
Chromosome Location
Chr.11: 119015712 - 119018347 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028.2 NP_001019.1 4 - 5 442 130
GenBank mRNA BC004294.1 - 1 - 1 470 130
BC004986.1 - 4 - 5 425 130

View Full Product Details