Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005516.5 | NP_005507.3 | 3 - 4 | 738 | 130 |
XM_017010807.1 | XP_016866296.1 | 2 - 3 | 750 | 130 | |
XM_017010808.1 | XP_016866297.1 | 2 - 3 | 750 | 130 | |
XM_017010809.1 | XP_016866298.1 | 3 - 4 | 635 | 130 | |
GenBank mRNA | AB209870.1 | - | 3 - 4 | 605 | 130 |
AJ293263.1 | - | 3 - 4 | 612 | 130 | |
AJ293264.1 | - | 3 - 4 | 612 | 130 | |
AK098434.1 | - | 3 - 4 | 635 | 130 | |
AK225048.1 | - | 3 - 4 | 632 | 130 | |
AK292391.1 | - | 3 - 4 | 690 | 130 | |
AK296822.1 | - | 2 - 3 | 762 | 130 | |
AK300906.1 | - | 3 - 4 | 558 | 130 | |
AK301125.1 | - | 3 - 4 | 413 | 130 | |
AK314411.1 | - | 3 - 4 | 632 | 130 | |
AK316327.1 | - | 3 - 4 | 563 | 130 | |
AY216681.1 | - | 3 - 4 | 612 | 130 | |
AY221103.1 | - | 3 - 4 | 612 | 130 | |
BC002578.2 | - | 3 - 4 | 626 | 130 | |
BC004297.1 | - | 3 - 4 | 610 | 130 | |
BC040479.1 | - | 3 - 4 | 632 | 130 | |
M20022.1 | - | 3 - 4 | 619 | 130 | |
M31183.1 | - | 3 - 4 | 584 | 130 |