Assay Design



Gene Transcripts

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
Chromosome Location
Chr.5: 158695915 - 159099762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290360.2 NP_001277289.1 13 - 14 1654 121
NM_001324101.1 NP_001311030.1 13 - 14 1654 121
NM_001324103.1 NP_001311032.1 13 - 14 1651 121
NM_001324106.1 NP_001311035.1 13 - 14 1627 121
NM_001324107.1 NP_001311036.1 13 - 14 1630 121
NM_001324108.1 NP_001311037.1 13 - 14 1627 121
NM_001324111.1 NP_001311040.1 13 - 14 1749 121
NM_024007.4 NP_076870.1 13 - 14 1651 121
NM_182708.2 NP_874367.1 12 - 13 1558 121
XM_017009192.1 XP_016864681.1 13 - 14 1654 121
XM_017009193.1 XP_016864682.1 13 - 14 1639 121
XM_017009194.1 XP_016864683.1 13 - 14 1630 121
XM_017009195.1 XP_016864684.1 13 - 14 1627 121
XM_017009196.1 XP_016864685.1 13 - 14 2589 121
XM_017009197.1 XP_016864686.1 13 - 14 2092 121
XM_017009198.1 XP_016864687.1 13 - 14 2011 121
XM_017009199.1 XP_016864688.1 8 - 9 2005 121
XM_017009200.1 XP_016864689.1 8 - 9 19141 121
XM_017009201.1 XP_016864690.1 8 - 9 19138 121
XM_017009204.1 XP_016864693.1 5 - 6 1531 121
GenBank mRNA AF208502.1 - 13 - 14 1353 121
AK289712.1 - 13 - 14 1638 121
AK303698.1 - 11 - 12 1280 121
AK304438.1 - 13 - 14 1357 121
BC038805.1 - 13 - 14 1662 121
BC041178.1 - 12 - 13 1485 121

View Full Product Details