Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020991.3 | NP_066271.1 | 5 - 5 | 803 | 82 |
NM_022644.3 | NP_072170.1 | 4 - 4 | 1056 | 82 | |
NM_022645.2 | NP_072171.1 | 3 - 3 | 518 | 82 | |
GenBank mRNA | BC022044.1 | - | 5 - 5 | 754 | 82 |
BC035965.1 | - | 5 - 5 | 750 | 82 | |
V00573.1 | - | 5 - 5 | 712 | 82 |