Assay Design



Gene Transcripts

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
Chromosome Location
Chr.X: 35919734 - 36385319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304548.1 NP_001291477.1 62 - 63 119
GenBank mRNA AI822140.1 - 2 - 3 406 119

View Full Product Details