Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291333.1 | NP_001278262.1 | 1 - 2 | 194 | 114 |
NM_004623.4 | NP_004614.3 | 1 - 2 | 194 | 114 | |
GenBank mRNA | AF063602.1 | - | 1 - 2 | 156 | 114 |
AF073887.1 | - | 1 - 2 | 140 | 114 | |
AK025524.1 | - | 1 - 2 | 194 | 114 | |
AK222562.1 | - | 1 - 2 | 147 | 114 | |
AK292426.1 | - | 1 - 2 | 155 | 114 | |
BC001276.1 | - | 1 - 2 | 133 | 114 | |
BT006817.1 | - | 1 - 2 | 107 | 114 |