Assay Design



Gene Transcripts

Gene Symbol
PSG2
Entrez Gene ID
5670
Gene Name
pregnancy specific beta-1-glycoprotein 2
Gene Aliases
CEA, PSBG2, PSG1
Chromosome Location
Chr.19: 43064210 - 43082741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031246.3 NP_112536.2 5 - 6 1223 103
GenBank mRNA BC022316.1 - 5 - 6 1150 103
M20882.1 - 5 - 6 1134 103
M37398.1 - 2 - 3 324 103

View Full Product Details