Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301107.1 | NP_001288036.1 | 5 - 6 | 593 | 113 |
NM_016053.3 | NP_057137.1 | 5 - 6 | 596 | 113 | |
XM_017019383.1 | XP_016874872.1 | 4 - 5 | 411 | 113 | |
GenBank mRNA | AF151874.1 | - | 5 - 6 | 541 | 113 |
AF155655.1 | - | 5 - 6 | 541 | 113 | |
AI739611.1 | - | 2 - 3 | 113 | ||
AK223339.1 | - | 5 - 6 | 532 | 113 | |
AK314970.1 | - | 5 - 6 | 514 | 113 | |
BC010889.1 | - | 6 - 7 | 467 | 113 | |
BX356987.2 | - | 5 - 6 | 548 | 113 | |
CR457171.1 | - | 5 - 6 | 434 | 113 |